Learn More
Cayman Chemical 25I-NBFhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1406610 MG
Encompass_Preferred
A 2C-I derivative that acts as a partial agonist of human 5-HT2A receptors (Ki = 0.26 nM; EC50 = 1.6 nM); intended for forensic and research applications
Catalog No. 50-295-5383