Learn More
Cayman Chemical 25I-NBOHhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1490910 MG
Encompass_Preferred
A derivative of 2C-I that acts as a potent agonist for the human serotonin 5-HT2A receptor (Ki = 0.061 nM); intended for research and forensic applications
Catalog No. 50-296-8909