Learn More
Avanti Polar Lipids 25MG PHOSPHATIDYLCHOLINE PWDR
Supplier:  Avanti Polar Lipids 850375P25MG
Encompass
18:1 PC (cis), 1,2-Dioleoyl-sn-Glycero-3-Phosphocholine (DOPC), Hygroscopic as Lyophilized White Powder, Molecular Weight: 786.15, Molecular Formula: C44H84NO8P
Catalog No. NC9168334