Learn More
Chemglass Life Sciences Flask, 25mL, Filtering, # 3 Rubber Stopper, With Hose Connection
SDP
Supplier:  Chemglass Life Sciences CG852225
Encompass_Preferred
Flask, 25mL, Filtering, # 3 Rubber Stopper, With Hose Connection. These small filtering flasks are recommended for microchemical use. All flasks have permanent white enamel marking spots. PLEASE NOTE: FOR LARGER CAPACITY FILTERING FLASKS, SEE CG-8514
Catalog No. NC1360307