Learn More
Bellco Glass, Inc. 25mL Pipet, Bulk Packed
SDP
Supplier:  Bellco Glass, Inc. 229245000
Encompass_Preferred
Convenient bulk packaging optionAll serological pipets are manufactured to the ASTM Standard Specification for Serological Pipets E-934Bi-directional (positive and negative) graduations are printed in bold easy-to-read inkPremium material barrier plugColor coded stripes and packaging enable quick volume identification Pack Description25 Bulk Packed/BagCase Quantity200SterileYesNon-pyrogenicYesPrimary ColorClearSecondary ColorRedPrimary MaterialPolystyreneMaximum Volume25mLGraduatedYesGraduation Increments (Subdivisions)2/10mLRNase/DNase FreeYes
Catalog No. 50-232-9954