Learn More
Pacer Global LLC 25ml Reagent Reservoir, Polystyrene, White, Sterile, 200 per Case
SDP
Supplier:  Pacer Global LLC MLSMRWS25/DEL
Encompass_Preferred
25mL Disposable Reagent Reservoir. Sterile. Polystyrene. White. Made from virgin grade resin. Lot Certified RNase/DNase-free. Non-Pyrogenic, Animal Free, Human DNA Free. Designed to accommodate single & multichannel pipettors. Stacking lugs prevent crushing & facilitate separation. Graduation marks on the interior walls. Pour-off spouts in each corner. 5/pack, 200/case.
Catalog No. NC1862157