Learn More
Genesee Scientific Corporation Genesee Scientific 25 mL Serological Pipets, Accurate Dispensing ±2%, Sterile, 25 mL
SDP
Supplier:  Genesee Scientific Corporation 12106
Encompass
The Genesee Scientific 25 mL Serological Pipets are designed for precise liquid handling in scientific applications. Made from optical grade polystyrene, they offer accuracy within ±2%, bi-directional graduations, and a patented filter for contamination protection. Sterilized through gamma irradiation, they are ideal for sensitive biological work. Their individual packaging promotes sterility, while color coding enhances identification, making them a reliable choice for laboratory workflows.
Catalog No. NC0635576