Learn More
HARVARD BIOSCIENCE INC 25MM COVER GLASS PK/100
Supplier:  HARVARD BIOSCIENCE INC 640705
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 11.Coverslip Glass, no.1 Thickness, 0.15mm or 0.006 in., 25mm
Catalog No. NC9299543