Learn More
Grainger Drum Thief Sampler, Sterilized 100ML
Supplier:  Grainger 8Y622
Encompass_Preferred
The Drum Thief Sampler Sterilized is a single-use, sterile sampling device designed for aseptic collection of liquid samples from drums and other containers. It features a polypropylene body with a polyethylene bulb that creates suction to draw the sample. The sampler has a capacity of 100mL and a minimum/maximum immersion depth of 2"/6". It can withstand temperatures up to 200°F. The Drum Thief is individually packaged to maintain sterility. This product enables safe, contamination-free sampling and is suitable for use in food, beverage, pharmaceutical, and other applications requiring sterile sampling.
Catalog No. NC2455775