Learn More
Cole-Parmer 25PK VALUE PLASTICS LUER FIT
Supplier:  Cole-Parmer 5012005
Encompass_Preferred
Cole-Parmer 3P Value Plastics Luer Fitting, Straight Adapter, Polypropylene, Male Luer Slip x Female Luer Thread; 25/Pk
Catalog No. NC3372422