Learn More
USA Scientific Inc 25-place upright freezer rack
Supplier:  USA Scientific Inc 26025500
Encompass
25-place (5 long x 5 high) upright freezer rack for 2" H boxes, 26 3/4" L x 5 1/2" W x 11" H, stainless steel
Catalog No. 50-274-4768