Learn More
Medchemexpress LLC (25R)-Spirost-4-ene-3,12-dione | 6875-60-1 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN78851MG
Encompass_Preferred
(25R)-Spirost-4-ene-3,12-dione is a natural product that inhibits neutrophil superoxide anion production and histamine release from mast cells. It is for research use only.
  • Natural product
  • Inhibits neutrophil superoxide anion production
  • Inhibits histamine release from mast cells
  • For research use only
Catalog No. 50-004-17841