Learn More
TARGETMOL CHEMICALS INC 2614W94 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T100895MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 2614W94 is a selective inhibitor of MAO-A with IC50 of 5 nM and Ki of 1.6 nM. purity: 99%
Catalog No. 50-258-1765