Learn More
Waters Corp WATERS CORPORATION Check Valve Cartridge, Replacement Kit, 2 pk, HPLC
Supplier:  Waters Corp WAT270941
Encompass_Preferred
These cartridges are integral to the 2690/2695 and 2790/2795 Separations Modules. They are critical to the overall performance of the Alliance™ system, ensuring long life, low internal volume, and quick response to flow changes.
  • Ensures long life.
  • Maintains low internal volume.
  • Provides quick response to flow changes.
  • Check valve type.
  • Designed for HPLC systems.
  • Two units per package.
  • Replaced as required.
Catalog No. 50-818-940