Learn More
Cayman Chemical 27-hydroxy Cholestenon 10mg
SDP
Supplier:  Cayman Chemical 2555010 MG
Encompass_Preferred
An oxysterol; levels are increased in Cyp46a-/- mouse brain; has been used as an intermediate in the synthesis of dafachronic acid ligands for the DAF-12 nuclear hormone receptor
Catalog No. 50-301-5731