Learn More
Medchemexpress LLC 27-Hydroxycholesterol | 20380-11-4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN2371
Encompass_Preferred
27-Hydroxycholesterol (27-OHC) is a selective estrogen receptor modulator and an agonist of the liver X receptor. It is an endogenous oxysterol that regulates intracellular cholesterol homeostasis and is intended for research use only.
  • Acts as an endogenous selective estrogen receptor modulator
  • Functions as an agonist of the liver X receptor
  • Positively regulates gene transcription and cell proliferation in breast cancer models
  • Increases cell proliferation in LNCaP and PC3 cells
  • Produced from cholesterol by monooxygenase CYP27A1
  • Levels can be modulated by statins or CYP27A1 inhibition
  • Important for LXR activation in response to cholesterol overload
Catalog No. 50-004-22029