Learn More
Waters Corp Waters Performance Maintenance Kit, 2707 Autosampler, HPLC System, Annual Replacement
Supplier:  Waters Corp 201000196
Encompass_Preferred
The Waters Performance Maintenance Kit for the 2707 Autosampler contains all the necessary parts to keep the 2707 Autosampler operating at peak performance on an annual basis.
  • Air needle (62 mm)
  • Standard needle assembly
  • Wash station with screws
  • Syringe valve
  • Buffer tubing (1000 μL)
  • Rotor seal
  • 500 μL syringe
  • Performance maintenance type
  • HPLC system compatibility
  • Annual replacement schedule
  • One unit per package
Catalog No. 50-791-585