Learn More
eMolecules​ 2712-78-9 | [Bis(trifluoroacetoxy)iodo]benzene | AA Blocks LLC | MFCD00009672 | 430.041 | C10H5F6IO4 | 0.000 | FC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)c1ccccc1 | 10g | 532057343
Supplier:  eMolecules​ 532057343
Encompass_Preferred
[Bis(trifluoroacetoxy)iodo]benzene | AA Blocks LLC | 2712-78-9 | MFCD00009672 | 430.041 | C10H5F6IO4 | 0.000 | FC(F)(F)C(=O)O[I](OC(=O)C(F)(F)F)c1ccccc1 | 10g | 532057343
Catalog No. 50-001-60741