Learn More
Polyscience 28 LITER S/S OPEN TANK
SDP
Supplier:  Polyscience T28SA1
Encompass_Preferred
Key SpecificationsDescription Stainless Steel Open Tank, 28LWorking Temperature Range °C Up to 150°Working Access (L x W x D) (cm) 44.5 x 24.1 x 20.3
Catalog No. NC2196805