Learn More
Chemglass Life Sciences Flask, Fernbach, 2800mL, 38mm OD Delong Top, 3 Side Baffles, Approx OD x Height (mm): 210 x 260
SDP
Supplier:  Chemglass Life Sciences CLS202216
Encompass_Preferred
Flask, Fernbach, 2800mL, 38mm OD Delong Top, 3 Side Baffles, Approx OD x Height (mm): 210 x 260. Flasks are designed for culturing organisms that require a large surface area to volume ratio. Flask has 3 baffles on lower outside edge to help oxygen transfer to the culture medium.
Catalog No. 50-121-5173