Learn More
Chemglass Life Sciences Sticky Mat, 11 x 11 in (280 x 280 mm) for Stackable Incubator Shakers
SDP
Supplier:  Chemglass Life Sciences CLS2545MAT
Encompass
Sticky mat provides a non-slip, adhesive surface to secure flasks and vessels on shaker platforms, reducing movement and breakage during incubation and shaking. Sized to fit standard stackable incubator shaker platforms and sold individually.
  • Secures vessels with a tacky surface during shaking.
  • Fits shaker platforms measuring 11 x 11 in (280 x 280 mm).
  • Reduces glassware movement and risk of breakage.
  • Easy to position and remove for cleaning or replacement.
  • Sold as a single mat per package.
Catalog No. NC2401326