Learn More
Beckman Coulter Multi-Disc tube adapter assembly, 28 mm polypropylene - fits JS-series rotors
Supplier:  Beckman Coulter 339103
Encompass
Beckman Coulter Multi-Disc Tube Adapter Assembly is a polypropylene adapter designed to hold 28 mm diameter tubes in compatible multi-disc centrifuge rotors. It adapts standard tubes for secure seating during high-speed centrifugation and is supplied individually.
  • Made from polypropylene for chemical resistance and durability.
  • Designed for 28 mm diameter tubes.
  • Supplied as a single adapter per pack.
  • Compatible with high-performance and high-capacity multi-disc rotors.
  • Simplifies secure seating and handling of tubes during centrifugation.
Catalog No. NC2932585