Learn More
ATCC 293; Embryonic Kidney; Genomic DNA, Human
Supplier:  ATCC CRL1573D
Encompass
High molecular weight genomic DNA isolated from the 293 human embryonal kidney cell line transformed with adenovirus 5 DNA (ATCC CRL-1573).
Catalog No. 50-238-2569