Learn More
Cell Biolabs Inc 293RTV Cell Line for Retroviral Packaging
Supplier:  Cell Biolabs Inc RV100
Encompass_Preferred
The 293RTV Cell Line is a permanent line established from primary embryonic human kidney cell transformed with human adenovirus type 5 DNA. The genes encoded by the E1 region of adenovirus (E1a and E1b) are expressed in these cells and participate in transactivation of viral promoters, allowing these cells to produce high levels of protein. The 293RTV Cell Line also expresses the SV40 large T antigen and a neomycin resistance gene, as does the parental 293T cell from which it is derived. However, 293RTV offers several advantages over the standard 293T cells: (1) Larger cell surface area resulting higher transfection and better yield of retrovirus; (2) Fast growth; and (3) Firm attachment to culture plates, which make it ideal for large scale culture and retroviral production.
Catalog No. NC9061083