Learn More
ATCC 293T, Embryonic Kidney Tissue, Frozen
Supplier:  ATCC CRL3216/DEL
Encompass
The 293T cell line, originally referred as 293tsA1609neo, is a highly transfectable derivative of human embryonic kidney 293 cells, and contains the SV40 T-antigen. This cell line is competent to replicate vectors carrying the SV40 region of replication. It gives high titers when used to produce retroviruses. It has been widely used for retroviral production, gene expression and protein production. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. NC0567849
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.