Learn More
Agilent Technologies Glyko 2-AB Oligomannose 5 (Man5), 100pmol
Supplier:  Agilent Technologies GKSB103
Encompass
N-Glycan labeled with 2-aminobenzamide (2-AB), for use as a qualitative standard in applications such as liquid chromatography (LC). Alternative glycan names: Man5, M5
Catalog No. NC9691419