Learn More
Cayman Chemical 2AmIno9HpyrIdo2 3bIndol 50mg
SDP
Supplier:  Cayman Chemical 4211450 MG
Encompass_Preferred
A mutagenic heterocyclic amine; has been found in cooked foods; mutagenic to S. typhimurium in the Ames test and induces SCEs in NL3 cells; induces hepatic and blood vessel tumor formation in mice at 0.08% in the diet; has been found in the urine of cigarette smokers
Catalog No. 50-294-5434