Learn More
Cayman Chemical 2ArachIdonyl Glycerold8 100ug
SDP
Supplier:  Cayman Chemical 362160100 UG
Encompass_Preferred
An internal standard for the quantification of 2-AG by GC- or LC-MS
Catalog No. 50-291-2011