Learn More
Cayman Chemical 2ArachIdonyl GlycRol ethR 5mg
SDP
Supplier:  Cayman Chemical 621655 MG
Encompass_Preferred
A potent and selective agonist of CB1 and GPR55 (EC50s = 10, 37, and >30,000 nM for CB1, GPR55, and CB2, respectively); binds to cannabinoid (CB) receptor 1 (Kis = 21.2 and >3,000 nM for CB1 and CB2, respectively, in a radioligand binding assay); displays the typical tetrad of CB activities in mice; more chemically stable than 2-AG with an endogenous half-life of hours rather than minutes; 10-fold less potent than 2-AG in eliciting typical CB1-mediated responses; reduces intraocular pressure in rabbits when administered at doses exceeding 50 UG per eye; increases dietary intake and enhances fat consumption in rats given access to both high-carbohydrate and high-fat diets
Catalog No. 50-289-3706