Learn More
Cayman Chemical 2Chloro4nItrophenyl aDma 500mg
SDP
Supplier:  Cayman Chemical 34459500 MG
Encompass_Preferred
A colorimetric α-amylase substrate; upon hydrolysis by α-amylases, 2-chloro-4-nitrophenol is released which can be quantified by colorimetric detection at 405 nm as a measure of enzyme activity; has been used to characterize the activity of human pancreatic or salivary amylase
Catalog No. 50-294-6777