Learn More
Cayman Chemical 2Chloro4nItrophenylaDgl 10mg
SDP
Supplier:  Cayman Chemical 1652710 MG
Encompass_Preferred
A conjugate of CNP and α-D-glucose, to be used as a chromogenic substrate for enzymes that target the linkage between CNP and α-D-glucose
Catalog No. 50-296-7944