Learn More
Cayman Chemical 2CLOroN6Cyclopentyladenos 10mg
SDP
Supplier:  Cayman Chemical 3647710 MG
Encompass_Preferred
An adenosine A1 receptor antagonist; selectively binds to adenosine A1 over adenosine A2 receptors (Kis = 0.4 and 3,900 nM, respectively); inhibits forskolin-induced adenylate cyclase activity in rat fat cell membranes (IC50 = 33 nM); increases the electroconvulsive threshold in a mouse model of MES-induced seizures at 0.25 and 0.5 mg/kg; reduces infarct size in a rabbit model of coronary artery occlusion-induced myocardial infarction at 0.125 mg/kg
Catalog No. 50-291-1417