Learn More
Chemglass Life Sciences Cryogenic Vial, Self-Standing, Internal Thread, 0.5 mL, Sterile, 2D Barcode (Racked, 10x10 Rack, 16 Racks/Case)
SDP
Supplier:  Chemglass Life Sciences CGN476308
Encompass_Preferred
0.5 mL self-standing cryogenic vials with internal thread and a permanently marked 2D barcode for reliable sample identification and automated scanning. Sterile, autoclavable polypropylene construction and a silicone O-ring cap provide secure sealing and integrity during low-temperature storage.
  • 0.5 mL capacity.
  • Internal thread.
  • Sterile (E-beam sterilized, SAL = 10^-6).
  • Permanent 2D barcode on each vial for scanning.
  • Self-standing, round-bottom design.
  • USP VI polypropylene; DNase/RNase and human DNA free.
  • Autoclavable and non-pyrogenic.
  • Clear graduations and white marking area.
  • Silicone O-ring screw cap for secure sealing.
  • Pack configuration: 10 x 10 per rack, 16 racks per case (1,600 vials/case).
Catalog No. 50-273-3381