Learn More
Chemglass Life Sciences Chemglass 2D-Barcode 0.5 mL Cryogenic Vial - Self-Standing, External Thread, Sterile, 50/pack
SDP
Supplier:  Chemglass Life Sciences CGN476316
Encompass_Preferred
The Chemglass 0.5 mL cryogenic vial features a permanent laser-etched 2D barcode for reliable sample identification and rack-based scanning. It is a self-standing, external-thread polypropylene vial sterilized by e-beam (SAL 10^-6) and designed for cryogenic storage and autoclave compatibility.
  • Permanent laser-etched 2D barcode on the vial bottom for rack-in-place scanning.
  • 0.5 mL capacity with clear graduations and white marking area for easy labeling.
  • USP VI polypropylene construction for chemical compatibility and low-temperature performance.
  • Sterilized by e-beam (SAL 10^-6); non-pyrogenic, DNase/RNase and human DNA free.
  • External thread with a silicone-free screw cap to minimize contamination risk.
  • Autoclavable and designed for cryogenic storage.
  • Packaged 50 per pack, 500 per box, 2,000 per case for bulk inventory needs.
Catalog No. 50-273-3373