Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, Self-Standing, Internal Thread, 1.2 mL, 2D Barcode (CGN-4763-23)
SDP
Supplier:  Chemglass Life Sciences CGN476323
Encompass_Preferred
This 1.2 mL self-standing cryogenic vial features an internal thread and a 2D barcode for reliable sample identification and tracking. Made from USP VI polypropylene, the sterile vial is autoclavable and suitable for low-temperature cryogenic storage. Bulk packaging options support high-throughput workflows.
  • Self-standing design for stable upright storage.
  • Internal thread profile for secure sealing with compatible caps.
  • Permanent 2D barcode for unambiguous sample identification and tracking.
  • Sterile packaging to minimize contamination risk.
  • USP VI polypropylene construction; autoclavable and suitable for cryogenic storage.
  • Available in bulk packs to support inventory and high-throughput workflows.
Catalog No. 50-273-3367