Learn More
Chemglass Life Sciences Chemglass 2D Barcode 2.0 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile, 50/pack
SDP
Supplier:  Chemglass Life Sciences CGN476324
Encompass_Preferred
Sterile, self-standing 2.0 mL cryogenic vial with internal thread and a permanent laser-etched 2D barcode for automated sample identification and reliable low-temperature storage. Constructed from USP VI polypropylene and supplied in bulk packs for laboratory and biobanking workflows.
  • Capacity: 2.0 mL.
  • Thread type: Internal thread.
  • Sterile: E-beam sterilized, SAL = 10^-6.
  • Material: USP VI polypropylene.
  • Unique marking: Permanent laser-etched 2D barcode on the bottom of each vial.
  • Pack quantities: 50 per pack, 500 per box, 2,000 per case.
  • Intended use: Cryogenic sample storage and automated identification.
Catalog No. 50-273-3366