Learn More
Chemglass Life Sciences 2D Barcode 3.0 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile, 50/pk
SDP
Supplier:  Chemglass Life Sciences CGN476325
Encompass_Preferred
Sterile 3.0 mL cryogenic vials with permanent 2D barcodes provide traceable sample identification and reliable low-temperature storage. The self-standing, round-bottom design and internal-thread screw cap reduce contamination risk while enabling easy rack compatibility.
  • Permanent 2D barcode laser-marked on the vial bottom for quick scanning
  • 3.0 mL volume with clear graduations and white marking area
  • USP VI polypropylene construction, autoclavable
  • Sterile, E-beam sterilized, SAL=10^-6
  • Self-standing, round-bottom design for stable storage
  • Special screw cap designed without silicone ring to reduce contamination risk
  • Packaged 50 per pack, 250 per box, 1,000 per case
  • DNase-, RNase-, and human DNA-free
Catalog No. 50-273-3365