Learn More
Chemglass Life Sciences Chemglass 2D Barcode 3.0 mL Cryogenic Vial, Self-Standing, Internal Thread, Sterile - 9 x 9 Rack, 648/Case
SDP
Supplier:  Chemglass Life Sciences CGN476313
Encompass_Preferred
3.0 mL cryogenic vial with a permanent laser-marked 2D barcode designed for reliable sample identification and high-throughput rack scanning. Self-standing construction and USP VI polypropylene material provide compatibility with low-temperature storage; supplied sterile and packaged in racks for efficient handling.
  • 3.0 mL capacity.
  • Self-standing, round-bottom design.
  • Internal thread.
  • USP VI polypropylene construction.
  • Sterile; DNase/RNase and human DNA free.
  • Sterilized by E-beam (SAL=10^-6).
  • Permanent laser-marked 2D barcode on the vial bottom for scanning without removal from the rack.
  • 100% barcode inspection to ensure readability and uniqueness.
  • Packaged in 9 x 9 racks; 8 racks per case (648 vials per case).
  • Designed for cryogenic/low-temperature storage.
Catalog No. 50-273-3376