Learn More
Chemglass Life Sciences Chemglass 2D Barcode 4.0 mL Cryogenic Vial, Self-Standing, External Thread, Sterile, 50/pk
SDP
Supplier:  Chemglass Life Sciences CGN476320
Encompass_Preferred
4.0 mL cryogenic vials featuring a permanent laser-engraved 2D barcode for reliable sample identification and rapid scanning. Self-standing external-thread polypropylene construction offers secure, low-temperature storage and compatibility with standard racks. Sterile and inspected vials are supplied in bulk pack formats to support laboratory workflows.
  • Permanent laser-engraved 2D barcode on the bottom for rack-friendly scanning.
  • Self-standing, external-thread design for secure closure and easy handling.
  • Made from USP VI polypropylene; non-pyrogenic and DNase/RNase and human DNA free.
  • Sterile (E-beam, SAL=10^-6) and 100% barcode inspection for traceability.
  • Autoclavable for additional sterilization needs.
  • Clear graduations and a white marking area for sample labeling.
  • Special screw cap without a silicone ring to reduce contamination risk.
Catalog No. 50-273-3370