Learn More
Chemglass Life Sciences Chemglass 4.0 mL Cryogenic Vial - Self-Standing, Internal Thread, Sterile, Permanent 2D Barcode, 50/pack
SDP
Supplier:  Chemglass Life Sciences CGN476326
Encompass_Preferred
This 4.0 mL cryogenic vial provides secure sample storage and traceability for low-temperature applications. The vial is self-standing with an internal thread and is supplied sterile; each vial is permanently marked with a unique 2D barcode for reliable identification and inventory management.
  • 4.0 mL nominal capacity.
  • Permanently marked 2D barcode for sample traceability.
  • Self-standing design for stable upright storage.
  • Internal thread with corresponding screw cap.
  • Sterile, E-beam sterilized (SAL 10^-6).
  • Manufactured from USP Class VI polypropylene.
  • Screw cap designed without a silicone ring to reduce contamination risk.
  • Available in multiples: 50 per pack, 250 per box, 1,000 per case.
  • Intended for cryogenic and low-temperature biobanking applications.
Catalog No. 50-273-3364