Learn More
Chemglass Life Sciences Chemglass Cryogenic Vial, 5.0 mL, Internal Thread, Sterile, Laser 2D Barcode, Racked (CGN-4763-15)
SDP
Supplier:  Chemglass Life Sciences CGN476315
Encompass_Preferred
Self-standing 5.0 mL sterile cryogenic vial with an internal-thread screw cap and a laser-etched 2D barcode on the bottom for automated tracking and rack scanning. Made from USP VI polypropylene and E-beam sterilized to SAL 10^-6, the vial and silicone O-ring cap provide secure sealing and bioclean properties suitable for cryogenic storage and biobanking.
  • Capacity: 5.0 mL.
  • Material: USP VI polypropylene.
  • Closure: internal thread with silicone O-ring screw cap.
  • Barcode: laser 2D barcode permanently marked on the bottom for scanning in racks.
  • Sterility: E-beam sterilized, SAL = 10^-6.
  • Bioclean: non-pyrogenic, DNase/RNase free, and human DNA free.
  • Packaging: available in racked and nest formats; typical packaging is 9 x 9 per rack, 6 racks per case (486 vials/case).
  • Shape: round bottom, self-standing.
Catalog No. 50-273-3374