Learn More
Cayman Chemical 2Deoxy3O Nmethylnthr 3.1mg
SDP
Supplier:  Cayman Chemical 384573.1 MG
Encompass_Preferred
A fluorescent derivative of ATP; inhibitor of B. anthracis edema factor AC toxin (Ki = 40 nM); decreases inositol phosphate formation in CHO-K1 cells expressing the human purinergic P2Y12 receptor at 100 µM
Catalog No. 50-291-6725