Learn More
Cayman Chemical 2DeoxyadenosIn5OdIphos 10mg
SDP
Supplier:  Cayman Chemical 3619510 MG
Encompass_Preferred
A nucleotide diphosphate; inhibits the polymerization of ADP and CDP by E. coli or P. fluorescens polynucleotide phosphorylase at 0.23 mM; levels are increased in erythrocytes isolated from patients with adenosine deaminase deficiency and SCID
Catalog No. 50-291-1869