Learn More
Cayman Chemical 2DeoxycytIdIn5OtrIphos 10mg
SDP
Supplier:  Cayman Chemical 3348410 MG
Encompass_Preferred
A nucleoside triphosphate; has commonly been used in PCR reaction mixtures as a substrate for DNA synthesis by DNA polymerases
Catalog No. 50-294-6514