Learn More
Cayman Chemical 2deoxyDGlucose6phosphate 10mg
SDP
Supplier:  Cayman Chemical 1714910 MG
Encompass_Preferred
A derivative of 2-DG that is produced in mammalian cells by the action of hexokinase; accumulation of 2-DG6P in cells can inhibit glycolysis, leading to cell death
Catalog No. 50-296-7857