Learn More
Medchemexpress LLC (2E)-2-[4-(1-benzimidazolyl)-3-methoxyphenyl]-3-nitro-2-propenoic acid. | 108883-90-5 | 98.7% | 285.25 g·mol⁻¹ | C14H11N3O4 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW1413741G
Encompass_Preferred
CB096 is a small-molecule research compound that inhibits repeat-associated non-AUG (RAN) translation by binding the 5′CGG/3′GGC internal loop of pathogenic r(G4C2)exp RNA, reducing production of toxic dipeptide repeats and mitigating RNA-mediated cellular dysfunctions. Intended for laboratory research use.
  • Inhibits RAN translation with reported IC50 of 20 μM in HEK293T cells.
  • Binds the 5′CGG/3′GGC internal loop of r(G4C2)exp RNA.
  • Reduces generation of toxic dipeptide repeats produced by RAN translation.
  • High purity (98.7%) suitable for research applications.
  • Available in multiple pack sizes including 1 G.
  • Molecular formula C14H11N3O4 and molecular weight 285.25 g·mol⁻¹.
Catalog No. 50-002-83012