Learn More
Medchemexpress LLC (2E)-3-[5-(4-tert-butylbenzenesulfonamido)-1-benzothiophen-2-yl]-N-hydroxyprop-2-enamide | 1215115-03-9 | 98.2% | 430.54 g·mol⁻¹ | C21H22N2O4S2 | 5 MG

Supplier: Medchemexpress LLC HY1346015MG
KH-3 is a small-molecule inhibitor of the RNA-binding protein HuR supplied as a solid research reagent for biochemical and cellular studies of HuR-mRNA interactions.
- HuR inhibitor suitable for biochemical and cell-based studies.
- High purity: 98.2% by RP-HPLC.
- Chemical formula C21H22N2O4S2.
- Molecular weight 430.54 g·mol⁻1.
- Appearance: off-white to light yellow solid.
- Structure information available (SMILES provided).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.