Learn More
Medchemexpress LLC (2E)-5-phenyl-1-(2-thienyl)-2-penten-1-one | 1239987-91-7 | 99.8% | 242.34 g/mol | C15H14OS | 5 MG

Supplier: Medchemexpress LLC HY1454685MG
GPR52 antagonist-1 is a research-grade small-molecule antagonist of the G protein-coupled receptor GPR52. In preclinical studies it reduces mutant huntingtin (mHTT) levels and promotes survival of striatal neurons; it has been used in vivo to rescue Huntington's disease-related phenotypes in mouse models.
- Potent GPR52 inhibition with IC50 0.63 μM.
- Used in vivo: 5 mg/kg, i.p., once daily for 4 weeks in HdhQ140 mice.
- High purity (99.8%) and solid appearance.
- Molecular formula C15H14OS; molecular weight 242.34 g/mol.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Intended for research use; follow institutional safety and handling guidelines.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.