Learn More
Cayman Chemical 2Ethylhexyl 4methoxycInama 50g
SDP
Supplier:  Cayman Chemical 3340150 G
Encompass_Preferred
A UVB blocker; topical application prevents UV-induced edema and formation of cyclobutane pyrimidine dimers in mouse skin; increases uterine weight in female rats (ED50 = 934 mg/kg per day); also induces bleaching of A. pulchra coral
Catalog No. 50-294-7334