Learn More
Cayman Chemical 2F-Peracetyl-Fucose 250mg
SDP
Supplier:  Cayman Chemical 30865250 MG
Encompass_Preferred
A protein fucosylation inhibitor; inhibits fucosylation of IgG1 monoclonal antibodies produced by CHO-DG44 cells at 50 µM; reduces intracellular GDP-fucose levels in CHO-DG44 cells; decreases intracellular fucose levels in A. thaliana seedlings at 100 µM; decreases root length in A. thaliana seedlings
Catalog No. 50-298-9898